Synonyms:
3,5-Dimethylanthranilic acid
D2553-1G
TCD2553-001GEA
104
USD
InStock
TCD2553-001G
2-Amino-3,5-dimethylbenzoic acid ≥98.0% (by titrimetric analysis)
2-Amino-3,5-dimethylbenzoic acid
CAS Number: 14438-32-5
MDL Number: MFCD00017099
Molecular Formula: C9H11NO2
Molecular Weight: 165.19
Purity/Analysis Method: 98.0% (T)
Form: Crystal
Other Product Names / Synonyms: 2-Amino-3,5-dimethylbenzoic Acid