D2717-5G
TCD2717-005GEA
144.13
USD
InStock
TCD2717-005G
Dibenzyl diselenide ≥95.0% (by titrimetric analysis)
Dibenzyl diselenide
CAS Number: 1482-82-2
MDL Number: MFCD00004767
Molecular Formula: C14H14Se2
Molecular Weight: 340.19
Purity/Analysis Method: 95.0% (T)
Form: Crystal
Melting point (°C): 93
Product Categories: Synthetic Organic Chemistry, Selenenylation
Other Product Names / Synonyms: Benzyl Diselenide