Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

EDTA magnesium disodium salt hydrate ≥99.0% (by titrimetric analysis)

Supplier: TCI America
Danger
Synonyms: Ethylenedinitrilotetra-acetic acid magensium disodium saltKestranal® 2S MgEthylenediaminetetra-acetic acid magensium disodium saltEDTA, Magnesium Derivative, Disodium SaltMagnesium disodium ethylenediaminetetraacetateEdetic acid magensium disodium salt

E0094-25G
TCE0094-025GEA 28.85 USD InStock
TCE0094-025G
EDTA magnesium disodium salt hydrate ≥99.0% (by titrimetric analysis)
EDTA magnesium disodium salt
Form: Crystal

Other Product Names / Synonyms: Magnesium Disodium Ethylenediaminetetraacetate


Formula: C₁₀H₁₂MgN₂Na₂O₈·xH₂O
MW: 358.5 g/mol (anhy)
Storage Temperature: Ambient
MDL Number: MFCD03613601
CAS Number: 14402-88-1
EINECS: 238-372-3
https://us-legacy.vwr.com/stibo/web/std.lang.all/76/90/28967690.jpg
Order Now
Specification Test Results
Purity/Analysis Method 99.0% (T)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty