Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Ethylhexanal ≥95.0%

Supplier: TCI America

E0125-25ML E0125-500ML
TCE0125-25MLEA 28.93 USD InStock
TCE0125-25ML TCE0125-500ML
2-Ethylhexanal ≥95.0%
2-Ethylhexanal
CAS Number: 123-05-7
MDL Number: MFCD00006987
Molecular Formula: C8H16O
Molecular Weight: 128.22
Purity/Analysis Method: 95.0% (GC)
Form: Clear Liquid
Boiling point (°C): 163
Melting point (°C): -85
Flash Point (°C): 46
Specific Gravity (20/20): 0.82

Other Product Names / Synonyms: 2-Ethylhexaldehyde, 2-Ethylhexyl Aldehyde, Octyl Aldehyde


Formula: C₈H₁₆O
MW: 128.21 g/mol
Boiling Pt: 163 °C
Melting Pt: –85 °C
Density: 0.82 (20 °C)
Flash Pt: 46 °C
Storage Temperature: Ambient
MDL Number: MFCD00006987
CAS Number: 123-05-7
EINECS: 204-596-5
UN: 1989
ADR: 3,III
https://us-legacy.vwr.com/stibo/web/std.lang.all/77/11/28967711.jpg
Order Now
Specification Test Results
Purity/Analysis Method 95.0% (GC)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty