Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Ethylmalonic acid ≥98.0% (by titrimetric analysis)

Supplier: TCI America

E0137-5G E0137-25G
TCE0137-5GEA 90.2 USD InStock
TCE0137-5G TCE0137-025G
Ethylmalonic acid ≥98.0% (by titrimetric analysis)
Ethylmalonic acid
CAS Number: 601-75-2
MDL Number: MFCD00002668
Molecular Formula: C5H8O4
Molecular Weight: 132.12
Purity/Analysis Method: 98.0% (T)
Form: Crystal
Melting point (°C): 112


Formula: C₅H₈O₄
Melting Pt: 112 °C
MDL Number: MFCD00002668
CAS Number: 601-75-2
EINECS: 210-007-2
https://us-legacy.vwr.com/stibo/web/std.lang.all/77/20/28967720.jpg
Order Now
Specification Test Results
Purity/Analysis Method 98.0% (T)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty