Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Eriochrome Black T

Supplier: TCI America
Warning
Synonyms: Sodium 3-hydroxy-4-(2-hydroxy-1-naphthylazo)-7-nitro-1-naphthalenesulphonate Solochrome Black T EBT

E0214-25G
TCE0214-025GEA 81.32 USD InStock
TCE0214-025G
Eriochrome Black T
Eriochrome Black T
CAS Number: 1787-61-7
MDL Number: MFCD00003935
Molecular Formula: C20H13N3O7S
Molecular Weight: 461.38
Form: Crystal
Color: Black

Other Product Names / Synonyms: 1-(1-Hydroxy-2-naphthylazo)-6-nitro-2-naphthol-4-sulfonic Acid Sodium Salt, Mordant Black 11


Formula: C₂₀H₁₂N₃NaO₇S
MW: 461.39 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00003935
CAS Number: 1787-61-7
EINECS: 217-250-3
UN: 3077
ADR: 9,III
Merck Index: 12,03709
https://us-legacy.vwr.com/stibo/web/std.lang.all/77/78/28967778.jpg
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty