Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

3-Fluorobenzaldehyde ≥97.0%

Supplier: TCI America
Warning

F0189-25ML
TCF0189-025MLEA 53.38 USD InStock
TCF0189-025ML
3-Fluorobenzaldehyde ≥97.0%
3-Fluorobenzaldehyde
CAS Number: 456-48-4
MDL Number: MFCD00003348
Molecular Formula: C7H5FO
Molecular Weight: 124.11
Purity/Analysis Method: 97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 173
Flash Point (°C): 56
Specific Gravity (20/20): 1.18


Formula: C₇H₅FO
MW: 124.11 g/mol
Boiling Pt: 173 °C
Density: 1.18 (20 °C)
Flash Pt: 56 °C
Storage Temperature: Ambient
MDL Number: MFCD00003348
CAS Number: 456-48-4
EINECS: 207-266-9
UN: 1989
ADR: 3,III
https://us-legacy.vwr.com/stibo/web/std.lang.all/87/97/28968797.jpg
Order Now
Specification Test Results
Purity/Analysis Method 97.0% (GC)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty