Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

(2-Fluorophenyl)acetonitrile ≥97.0%

Supplier: TCI America
Synonyms: 2-Fluorobenzyl cyanide2-(2-Fluorophenyl)acetonitrile

F0257-25G
TCF0257-025GEA 61 USD InStock
TCF0257-025G
(2-Fluorophenyl)acetonitrile ≥97.0%
(2-Fluorophenyl)acetonitrile
CAS Number: 326-62-5
MDL Number: MFCD00001897
Molecular Formula: C8H6FN
Molecular Weight: 135.14
Purity/Analysis Method: 97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 235
Flash Point (°C): 43
Specific Gravity (20/20): 1.14

Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks
Other Product Names / Synonyms: 2-Fluorophenylacetonitrile


Formula: C₈H₆FN
MW: 135.14 g/mol
Boiling Pt: 235 °C
Density: 1.14 (20 °C)
Flash Pt: 43 °C
MDL Number: MFCD00001897
CAS Number: 326-62-5
EINECS: 206-313-0
UN: 3276
ADR: 6.1,III
https://us-legacy.vwr.com/stibo/web/std.lang.all/88/56/28968856.jpg
Order Now
Specification Test Results
Purity/Analysis Method 97.0% (GC)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty