Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Fluorobenzophenone

Supplier: TCI America
Warning
Synonyms: o-Fluorobenzophenone

F0397-5G
TCF0397-005GEA 59 USD InStock
TCF0397-005G
2-Fluorobenzophenone
2-Fluorobenzophenone
CAS Number: 342-24-5
MDL Number: MFCD00000318
Molecular Formula: C13H9FO
Molecular Weight: 200.21
Form: Clear Liquid
Color: Pale Yellow
Boiling point (°C): 190
Flash Point (°C): 110
Specific Gravity (20/20): 1.19


Formula: C₁₃H₉FO
MW: 200.21 g/mol
Boiling Pt: 190 °C
Density: 1.19 (20 °C)
Flash Pt: 110 °C
MDL Number: MFCD00000318
CAS Number: 342-24-5
EINECS: 206-440-1
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty