Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Hydrazinium hydroxide ≥79%

Supplier: TCI America
Synonyms: Hydrazine hydrate Hydrazine monohydrate

H0204-500G H0204-25G
TCH0204-500GEA 66.43 USD InStock
TCH0204-500G TCH0204-25G
Hydrazinium hydroxide ≥79%
Hydrazinium hydroxide


Formula: H₆N₂O
MW: 50.06 g/mol
Boiling Pt: 119 °C
Melting Pt: –52 °C
Density: 1.03 (20 °C)
Flash Pt: 75 °C
MDL Number: MFCD00149931
CAS Number: 7803-57-8
EINECS: 206-114-9
UN: 2030
ADR: 8,II
https://us-legacy.vwr.com/stibo/web/std.lang.all/02/49/28970249.jpg
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty