Synonyms:
Hexanen
, Hexane
, Hexyl hydride
H0394-100ML
TCH0394-100MLEA
105.56
USD
InStock
TCH0394-100ML
n-Hexane ≥96.0%
n-Hexane
[for Spectrophotometry]
CAS Number: 110-54-3
MDL Number: MFCD00009520
Molecular Formula: C6H14
Molecular Weight: 86.18
Purity/Analysis Method: 96.0% (GC)
Form: Clear Liquid
Boiling point (°C): 68
Melting point (°C): -95
Flash Point (°C): -22
Specific Gravity (20/20): 0.66