Synonyms:
(±)-γ-Caprolactone
, (±)-Hexan-4-olide
, γ-Caprolactone
, 5-Ethyldihydrofuran-2(3H)-one
H0514-25ML
TCH0514-025MLEA
34.61
USD
InStock
TCH0514-025ML
(±)-γ-Hexalactone ≥98.0%
(±)-γ-Hexalactone
CAS Number: 695-06-7
MDL Number: MFCD00005401
Molecular Formula: C6H10O2
Molecular Weight: 114.14
Purity/Analysis Method: 98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 220
Flash Point (°C): 98
Specific Gravity (20/20): 1.03
Product Categories: Materials Chemistry, Monomers, Bioscience, Lipids
Other Product Names / Synonyms: gamma-Caprolactone