H0631-25G
TCH0631-025GEA
228.38
USD
InStock
TCH0631-025G
2-Amino-5-hydroxybenzoic acid ≥98.0% (by HPLC, titration analysis)
2-Amino-5-hydroxybenzoic acid
CAS Number: 394-31-0
MDL Number: MFCD00007870
Molecular Formula: C7H7NO3
Molecular Weight: 153.14
Purity/Analysis Method: 98.0% (HPLC,T)
Form: Crystal
Other Product Names / Synonyms: 2-Amino-5-hydroxybenzoic Acid