Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-Hexylbenzoic acid ≥98.0% (by GC, titration analysis)

Supplier: TCI America

H0829-5G
TCH0829-005GEA 156.2 USD InStock
TCH0829-005G
4-Hexylbenzoic acid ≥98.0% (by GC, titration analysis)
4-Hexylbenzoic acid
CAS Number: 21643-38-9
MDL Number: MFCD00013997
Molecular Formula: C13H18O2
Molecular Weight: 206.29
Purity/Analysis Method: 98.0% (GC,T)
Form: Crystal


Formula: C₁₃H₁₈O₂
MW: 206.28 g/mol
Boiling Pt: 330 °C (1013 hPa)
Melting Pt: 97 °C
Storage Temperature: Ambient
MDL Number: MFCD00013997
CAS Number: 21643-38-9
EINECS: 244-490-6
https://us-legacy.vwr.com/stibo/web/std.lang.all/08/54/28970854.jpg
Order Now
Specification Test Results
Purity/Analysis Method 98.0% (GC,T)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty