Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

1-Acetyl-2-naphthol ≥98.0%

Supplier: TCI America
Synonyms: 1-(2-Hydroxy-1-naphthyl)ethan-1-one2'-Hydroxy-1'-acetonaphthone1-Acetyl-2-hydroxynaphthalene1-(2-Hydroxynaphthalen-1-yl)ethanone2-Hydroxy-1-acetonaphthone

H0936-25G
TCH0936-025GEA 63.95 USD InStock
TCH0936-025G
1-Acetyl-2-naphthol ≥98.0%
1-Acetyl-2-naphthol
CAS Number: 574-19-6
MDL Number: MFCD00012131
Molecular Formula: C12H10O2
Molecular Weight: 186.21
Purity/Analysis Method: 98.0% (GC)
Form: Crystal
Boiling point (°C): 178
Melting point (°C): 65

Other Product Names / Synonyms: 1-Acetyl-2-hydroxynaphthalene, 1-Acetyl-2-naphthol


Formula: C₁₂H₁₀O₂
MW: 186.21 g/mol
Boiling Pt: 178 °C
Melting Pt: 65 °C
MDL Number: MFCD00012131
CAS Number: 574-19-6
EINECS: 209-369-4
https://us-legacy.vwr.com/stibo/web/std.lang.all/09/54/28970954.jpg
Order Now
Specification Test Results
Purity/Analysis Method 98.0% (GC)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty