Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Isobutyric acid ≥99.0%

Supplier: TCI America
Warning
Synonyms: 2-Methylpropionic acid2-Methylpropanoic acid

I0103-25ML I0103-500ML
TCI0103-500MLEA 30.02 USD InStock
TCI0103-500ML TCI0103-25ML
Isobutyric acid ≥99.0%
Isobutyric acid
CAS Number: 79-31-2
MDL Number: MFCD00002658
Molecular Formula: C4H8O2
Molecular Weight: 88.11
Purity/Analysis Method: 99.0% (GC)
Form: Clear Liquid
Boiling point (°C): 155
Melting point (°C): -47
Flash Point (°C): 56
Specific Gravity (20/20): 0.95


Formula: C₄H₈O₂
MW: 88.11 g/mol
Boiling Pt: 155 °C
Melting Pt: –47 °C
Density: 0.95 (20 °C)
Flash Pt: 56 °C
Storage Temperature: Ambient
MDL Number: MFCD00002658
CAS Number: 79-31-2
EINECS: 201-195-7
UN: 2529
ADR: 3,III
Merck Index: 13,05172
https://us-legacy.vwr.com/stibo/web/std.lang.all/18/88/28971888.jpg
Order Now
Specification Test Results
Purity/Analysis Method 99.0% (GC)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty