Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Ethyl isobutyrate ≥99.0%

Supplier: TCI America
Synonyms: 2-Methylpropionic acid ethyl ester

I0105-500ML I0105-25ML
TCI0105-500MLEA 60.88 USD InStock
TCI0105-500ML TCI0105-25ML
Ethyl isobutyrate ≥99.0%
Ethyl isobutyrate
CAS Number: 97-62-1
MDL Number: MFCD00009165
Molecular Formula: C6H12O2
Molecular Weight: 116.16
Purity/Analysis Method: 99.0% (GC)
Form: Clear Liquid
Boiling point (°C): 110
Melting point (°C): -88
Flash Point (°C): 15
Specific Gravity (20/20): 0.87

Other Product Names / Synonyms: Isobutyric Acid Ethyl Ester


Formula: C₆H₁₂O₂
MW: 116.16 g/mol
Boiling Pt: 110 °C
Melting Pt: –88 °C
Density: 0.87 (20 °C)
Flash Pt: 15 °C
MDL Number: MFCD00009165
CAS Number: 97-62-1
EINECS: 202-595-4
UN: 2385
ADR: 3,II
Merck Index: 13,03847
https://us-legacy.vwr.com/stibo/web/std.lang.all/18/89/28971889.jpg
Order Now
Specification Test Results
Purity/Analysis Method 99.0% (GC)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty