Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Isovaleraldehyde ≥98.0%

Supplier: TCI America
Synonyms: 3-Methylbutyraldehyde

I0192-25ML I0192-500ML I0192-100ML
TCI0192-25MLEA 21.21 USD InStock
TCI0192-25ML TCI0192-100ML TCI0192-500ML
Isovaleraldehyde ≥98.0%
Isovaleraldehyde
CAS Number: 590-86-3
MDL Number: MFCD00007014
Molecular Formula: C5H10O
Molecular Weight: 86.13
Purity/Analysis Method: 98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 93
Flash Point (°C): 3
Specific Gravity (20/20): 0.80

Other Product Names / Synonyms: 3-Methylbutanal


Formula: C₅H₁₀O
MW: 86.13 g/mol
Boiling Pt: 93 °C
Melting Pt: –60 °C
Density: 0.80 (20 °C)
Flash Pt: 3 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00007014
CAS Number: 590-86-3
EINECS: 209-691-5
UN: 2058
ADR: 3,II
Merck Index: 13,05250
https://us-legacy.vwr.com/stibo/web/std.lang.all/19/83/28971983.jpg
Order Now
Specification Test Results
Purity/Analysis Method 98.0% (GC)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty