Synonyms:
(S)-(+)-2,6-Diaminocaproic acid dihydrochloride (S)-(+)-2,6-Diaminohexanoic acid dihydrochloride (S)-(+)-Lysine dihydrochloride H-Lys-OH.2HCl
L0131-25G
TCL0131-025GEA
32.46
USD
InStock
TCL0131-025G
L(+)-Lysine dihydrochloride ≥98.0% (by titrimetric analysis)
L(+)-Lysine dihydrochloride
CAS Number: 657-26-1
MDL Number: MFCD00066389
Molecular Formula: C6H14N2O2
Molecular Weight: 219.11
Purity/Analysis Method: 98.0% (T)
Form: Crystal
Specific rotation [a]20/D: 15 deg (C=2, H2O)
Product Categories: Synthetic Organic Chemistry, Optical Resolving Agents, Bioscience, Proteinogenic Amino Acids
Other Product Names / Synonyms: H-Lys-OH.2HCl