Synonyms:
(S)-(+)-2-Hydroxypropanoic acid (S)-(+)-2-Hydroxypropionic acid (S)-(+)-Lactic acid
L0165-25G
L0165-500G
TCL0165-25GEA
41.18
USD
InStock
TCL0165-25G
TCL0165-500G
L(+)-Lactic acid ≥85.0% (by titrimetric analysis)
L(+)-Lactic acid
CAS Number: 79-33-4
MDL Number: MFCD00064266
Molecular Formula: C3H6O3
Molecular Weight: 90.08
Purity/Analysis Method: 85.0% (T)
Form: Clear Liquid
Flash Point (°C): 110
Specific Gravity (20/20): 1.21
Specific rotation [a]20/D: -13 deg (C=2.5, 1.5mol/L NaOH sol.)
Product Categories: Bioscience, Buffer-related Reagents
Other Product Names / Synonyms: (S)-2-Hydroxypropionic Acid