Synonyms:
2-Methoxytoluene
, o-Cresol methyll ether
, 2-Methylanisole
, 1-Methoxy-2-methylbenzene
M0150-500ML
M0150-25ML
TCM0150-500MLEA
242.51
USD
InStock
TCM0150-500ML
TCM0150-025ML
2-Methylanisole ≥98.0%
2-Methylanisole
CAS Number: 578-58-5
MDL Number: MFCD00008373
Molecular Formula: C8H10O
Molecular Weight: 122.17
Purity/Analysis Method: 98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 171
Flash Point (°C): 52
Specific Gravity (20/20): 0.98
Other Product Names / Synonyms: o-Cresol Methyl Ether, 2-Methylanisole