Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

(±)-3-Methyl-2-butanol ≥98.0%

Supplier: TCI America
Danger
Synonyms: DL-3-Methyl-2-butanol

M0171-25ML M0171-500ML
TCM0171-500MLEA 248.56 USD InStock
TCM0171-500ML TCM0171-25ML
(±)-3-Methyl-2-butanol ≥98.0%
(±)-3-Methyl-2-butanol
CAS Number: 598-75-4
MDL Number: MFCD00004527
Molecular Formula: C5H12O
Molecular Weight: 88.15
Purity/Analysis Method: 98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 112
Flash Point (°C): 39
Specific Gravity (20/20): 0.82

Other Product Names / Synonyms: sec-Isoamyl Alcohol, Isopropylmethylcarbinol, 2-Methyl-3-butanol


Formula: C₅H₁₂O
MW: 88.15 g/mol
Boiling Pt: 112 °C
Melting Pt: –117 °C
Density: 0.82 (20 °C)
Flash Pt: 39 °C
Storage Temperature: Ambient
MDL Number: MFCD00004527
CAS Number: 598-75-4
EINECS: 209-950-2
UN: 1105
ADR: 3,III
Merck Index: 12,06110
https://us-legacy.vwr.com/stibo/web/std.lang.all/35/02/28973502.jpg
Order Now
Specification Test Results
Purity/Analysis Method 98.0% (GC)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty