Synonyms:
α,4-Dimethylbenzyl alcohol
, 4-(1-Hydroxyethyl)toluene
, 1-(p-Tolyl)ethanol
, 1-(4-Methylphenyl)ethanol
, 4-Methylphenyl methyl carbinol
M0448-25ML
TCM0448-025MLEA
66.99
USD
InStock
TCM0448-025ML
p,α-Dimethylbenzyl alcohol ≥97.0%
p,α-Dimethylbenzyl alcohol
CAS Number: 536-50-5
MDL Number: MFCD00016860
Molecular Formula: C9H12O
Molecular Weight: 136.19
Purity/Analysis Method: 97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 220
Flash Point (°C): 107
Specific Gravity (20/20): 0.99
Other Product Names / Synonyms: alpha,4-Dimethylbenzyl Alcohol, 4-(1-Hydroxyethyl)toluene, Methyl-p-tolylcarbinol