Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Methyltrimethoxysilane ≥98.0%

Supplier: TCI America
Synonyms: Trimethoxy(methyl)silane

M0660-100ML M0660-25ML M0660-500ML
TCM0660-500MLEA 68.99 USD InStock
TCM0660-500ML TCM0660-100ML TCM0660-25ML
Methyltrimethoxysilane ≥98.0%
Methyltrimethoxysilane
CAS Number: 1185-55-3
MDL Number: MFCD00008342
Molecular Formula: C4H12O3Si
Molecular Weight: 136.22
Purity/Analysis Method: 98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 103
Flash Point (°C): 13
Specific Gravity (20/20): 0.96


Formula: C₄H₁₂O₃Si
Boiling Pt: 103 °C
Melting Pt: 11 °C
Density: 0.96 (20 °C)
Flash Pt: 13 °C
Storage Temperature: Ambient
MDL Number: MFCD00008342
CAS Number: 1185-55-3
EINECS: 214-685-0
https://us-legacy.vwr.com/stibo/web/std.lang.all/41/23/28974123.jpg
Order Now
Specification Test Results
Purity/Analysis Method 98.0% (GC)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty