Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Methoxyacetic acid ≥96.0% (by titrimetric analysis)

Supplier: TCI America
Synonyms: 2-Methoxyacetic acid

M0827-25G M0827-500G
TCM0827-500GEA 107.56 USD InStock
TCM0827-500G TCM0827-25G
Methoxyacetic acid ≥96.0% (by titrimetric analysis)
Methoxyacetic acid
CAS Number: 625-45-6
MDL Number: MFCD00004308
Molecular Formula: C3H6O3
Molecular Weight: 90.08
Purity/Analysis Method: 96.0% (T)
Form: Clear Liquid
Boiling point (°C): 204
Flash Point (°C): 125
Specific Gravity (20/20): 1.18


Formula: C₃H₆O₃
MW: 90.08 g/mol
Boiling Pt: 204 °C
Melting Pt: 7 °C
Density: 1.18 (20 °C)
Flash Pt: 125 °C
MDL Number: MFCD00004308
CAS Number: 625-45-6
EINECS: 210-894-6
UN: 3265
ADR: 8,II
https://us-legacy.vwr.com/stibo/web/std.lang.all/42/97/28974297.jpg
Order Now
Specification Test Results
Purity/Analysis Method 96.0% (T)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty