Synonyms:
(2-Methylphenyl)methanol
, 2-(Hydroxymethyl)toluole
M0887-25G
TCM0887-025GEA
58.87
USD
InStock
TCM0887-025G
2-Methylbenzyl alcohol ≥97.0%
2-Methylbenzyl alcohol
CAS Number: 89-95-2
MDL Number: MFCD00004622
Molecular Formula: C8H10O
Molecular Weight: 122.17
Purity/Analysis Method: 97.0% (GC)
Form: Crystal
Boiling point (°C): 219
Melting point (°C): 36
Flash Point (°C): 104
Other Product Names / Synonyms: alpha-Hydroxy-o-xylene