Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

N-Methylmorpholine-N-oxide 50% ∼4.8 mol/L in water

Supplier: TCI America
Synonyms: 4-Methylmorpholine 4-oxide4-Methylmorpholine N-Oxide

M0981-25ML M0981-500ML
TCM0981-500MLEA 58.06 USD InStock
TCM0981-500ML TCM0981-025ML
N-Methylmorpholine-N-oxide 50% ∼4.8 mol/L in water
N-Methylmorpholine N-oxide
CAS Number: 7529-22-8
MDL Number: MFCD00005947
Molecular Formula: C5H11NO2
Molecular Weight: 117.15
Form: Clear Liquid
Color: Pale Yellow
Specific Gravity (20/20): 1.12

Other Product Names / Synonyms: NMO


Formula: C₅H₁₁NO₂
MW: 117.15 g/mol
Density: 1.12 (20 °C)
MDL Number: MFCD00005947
CAS Number: 7529-22-8
EINECS: 231-391-8
UN: 3082
ADR: 9,III
https://us-legacy.vwr.com/stibo/web/std.lang.all/45/66/28974566.jpg
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty