N0019-100ML
TCN0019-100MLEA
84.85
USD
InStock
TCN0019-100ML
Nitromethane ≥98.0%
Nitromethane
[for Spectrophotometry]
CAS Number: 75-52-5
MDL Number: MFCD00007400
Molecular Formula: CH3NO2
Molecular Weight: 61.04
Purity/Analysis Method: 98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 101
Melting point (°C): -29
Flash Point (°C): 35
Specific Gravity (20/20): 1.14
Lambda max.: 380 nm (H2O)