Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Methyl-4-nitrobenzoate ≥98.0%

Supplier: TCI America
Synonyms: 4-Nitrobenzoic acid emthyl esterMethyl 4-nitrobenzoate

N0159-25G
TCN0159-025GEA 131.9 USD InStock
TCN0159-025G
Methyl-4-nitrobenzoate ≥98.0%
Methyl 4-nitrobenzoate
CAS Number: 619-50-1
MDL Number: MFCD00007350
Molecular Formula: C8H7NO4
Molecular Weight: 181.15
Purity/Analysis Method: 98.0% (GC)
Form: Crystal
Melting point (°C): 96

Other Product Names / Synonyms: 4-Nitrobenzoic Acid Methyl Ester


Formula: C₈H₇NO₄
MW: 181.15 g/mol
Melting Pt: 96 °C
Storage Temperature: Ambient
MDL Number: MFCD00007350
CAS Number: 619-50-1
EINECS: 210-599-2
https://us-legacy.vwr.com/stibo/web/std.lang.all/92/55/28989255.jpg
Order Now
Specification Test Results
Purity/Analysis Method 98.0% (GC)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty