N0214-1G
TCN0214-001GEA
86.28
USD
InStock
TCN0214-001G
5-Nitro-1,10-phenanthroline ≥98.0% (by HPLC, titration analysis)
5-Nitro-1,10-phenanthroline
CAS Number: 4199-88-6
MDL Number: MFCD00004981
Molecular Formula: C12H7N3O2
Molecular Weight: 225.21
Purity/Analysis Method: 98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 202