Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

5-Nitrosalicylic acid ≥98.0% (by GC, titration analysis)

Supplier: TCI America
Warning
Synonyms: 2-Hydroxy-5-nitrobenzoic acid

N0257-25G N0257-250G
TCN0257-025GEA 52.78 USD InStock
TCN0257-025G TCN0257-250G
5-Nitrosalicylic acid ≥98.0% (by GC, titration analysis)
5-Nitrosalicylic acid
CAS Number: 96-97-9
MDL Number: MFCD00007338
Molecular Formula: C7H5NO5
Molecular Weight: 183.12
Purity/Analysis Method: 98.0% (GC,T)
Form: Crystal
Melting point (°C): 236

Other Product Names / Synonyms: 2-Hydroxy-5-nitrobenzoic Acid


Formula: C₇H₅NO₅
MW: 183.12 g/mol
Melting Pt: 236 °C
Density: 1.65 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00007338
CAS Number: 96-97-9
EINECS: 202-548-8
Merck Index: 12,06730
https://us-legacy.vwr.com/stibo/web/std.lang.all/93/41/28989341.jpg
Order Now
Specification Test Results
Purity/Analysis Method 98.0% (GC,T)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty