Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

DL-Norvaline ≥98.0% (by titrimetric analysis)

Supplier: TCI America
Synonyms: (±)-Norvaline H-DL-Nva-OH

N0304-25G
TCN0304-025GEA 25.38 USD InStock
TCN0304-025G
DL-Norvaline ≥98.0% (by titrimetric analysis)
DL-Norvaline
CAS Number: 760-78-1
MDL Number: MFCD00064420
Molecular Formula: C5H11NO2
Molecular Weight: 117.15
Purity/Analysis Method: 98.0% (T)
Form: Crystal

Product Categories: Bioscience, non-Proteinogenic Amino Acids
Other Product Names / Synonyms: 2-Aminovaleric Acid


Formula: C₅H₁₁NO₂
MW: 117.15 g/mol
Boiling Pt: 223 °C (1013 hPa)
Melting Pt: > 300 °C
Storage Temperature: Ambient
MDL Number: MFCD00064420
CAS Number: 760-78-1
EINECS: 212-082-7
Merck Index: 12,06813
https://us-legacy.vwr.com/stibo/web/std.lang.all/93/82/28989382.jpg
Order Now
Specification Test Results
Purity/Analysis Method 98.0% (T)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty