Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

5-Nitro-1H-indazole ≥98.0%

Supplier: TCI America
Synonyms: 5-NINA5-Nitro-1H-indazole5-Nitroindazole

N0398-25G
TCN0398-025GEA 58.87 USD InStock
TCN0398-025G
5-Nitro-1H-indazole ≥98.0%
5-Nitro-1H-indazole
CAS Number: 5401-94-5
MDL Number: MFCD00005693
Molecular Formula: C7H5N3O2
Molecular Weight: 163.14
Purity/Analysis Method: 98.0% (GC)
Form: Crystal
Melting point (°C): 210


Formula: C₇H₅N₃O₂
MW: 163.14 g/mol
Melting Pt: 210 °C
MDL Number: MFCD00005693
CAS Number: 5401-94-5
EINECS: 226-451-5
https://us-legacy.vwr.com/stibo/web/std.lang.all/94/84/28989484.jpg
Order Now
Specification Test Results
Purity/Analysis Method 98.0% (GC)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty