Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Orcein

Supplier: TCI America
Synonyms: Natural Red 28

O0061-1G
TCO0061-001GEA 25.38 USD InStock
TCO0061-001G
Orcein
Orcein
CAS Number: 1400-62-0
MDL Number: MFCD00062310
Form: Crystal
Color: Deep Yellow Red

Product Categories: Bioscience, Stains & Dyes


MDL Number: MFCD00062310
CAS Number: 1400-62-0
EINECS: 215-750-6
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty