O0118-100ML
TCO0118-100MLEA
277.37
USD
InStock
TCO0118-100ML
n-Octane ≥98.5%
n-Octane
[for Spectrophotometry]
CAS Number: 111-65-9
MDL Number: MFCD00009556
Molecular Formula: C8H18
Molecular Weight: 114.23
Purity/Analysis Method: 98.5% (GC)
Form: Clear Liquid
Boiling point (°C): 125
Melting point (°C): -57
Flash Point (°C): 14
Specific Gravity (20/20): 0.70