Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

DL-α-Phenylalanine ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America
Synonyms: 3-Phenyl-DL-alanine DL-Phenylalanine H-DL-Phe-OH

P0136-250G P0136-25G
TCP0136-025GEA 33.5 USD InStock
TCP0136-025G TCP0136-250G
DL-α-Phenylalanine ≥98.0% (by HPLC, titration analysis)
DL-α-Phenylalanine
CAS Number: 150-30-1
MDL Number: MFCD00064225
Molecular Formula: C9H11NO2
Molecular Weight: 165.19
Purity/Analysis Method: 98.0% (HPLC,T)
Form: Crystal

Product Categories: Bioscience, Proteinogenic Amino Acids
Other Product Names / Synonyms: H-DL-Phe-OH


Formula: C₉H₁₁NO₂
MW: 165.19 g/mol
Boiling Pt: 313 °C (1013 hPa)
Melting Pt: 173 °C
MDL Number: MFCD00064225
CAS Number: 150-30-1
EINECS: 205-756-7
Merck Index: 12,07425
https://us-legacy.vwr.com/stibo/web/std.lang.all/08/46/28990846.jpg
Order Now
Specification Test Results
Purity/Analysis Method 98.0% (HPLC,T)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty