Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Phenylarsine oxide

Supplier: TCI America
Synonyms: OxophenylarsineArzenePAO

P0140-5G
TCP0140-005GEA 90.34 USD InStock
TCP0140-005G
Phenylarsine oxide
Phenylarsine oxide
CAS Number: 637-03-6
MDL Number: MFCD00001990
Molecular Formula: C6H5AsO
Molecular Weight: 168.03
Form: Crystal
Color: White


Formula: C₆H₅AsO
MW: 168.02 g/mol
MDL Number: MFCD00001990
CAS Number: 637-03-6
EINECS: 211-275-3
UN: 3465
ADR: 6.1,III
https://us-legacy.vwr.com/stibo/web/std.lang.all/08/53/28990853.jpg
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty