Synonyms:
P,P-Dichlorophenylphosphine
, Phenyldichlorophosphine
P0207-500ML
P0207-25ML
TCP0207-500MLEA
140.87
USD
InStock
TCP0207-500ML
TCP0207-25ML
Dichloro(phenyl)phosphine ≥98.0% (by titrimetric analysis)
Dichloro(phenyl)phosphine
CAS Number: 644-97-3
MDL Number: MFCD00000528
Molecular Formula: C6H5Cl2P
Molecular Weight: 178.98
Purity/Analysis Method: 98.0% (T)
Form: Clear Liquid
Boiling point (°C): 59
Flash Point (°C): 102
Specific Gravity (20/20): 1.32
Other Product Names / Synonyms: Benzenephosphonous Dichloride, Phenylphosphonous Dichloride