Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

N,N,N-Trimethylanilinium chloride ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America
Danger
Synonyms: Trimethylphenylammonium chloridePhenyltrimethylammonium chloride

P0244-25G
TCP0244-025GEA 22.33 USD InStock
TCP0244-025G
N,N,N-Trimethylanilinium chloride ≥98.0% (by HPLC, titration analysis)
N,N,N-Trimethylanilinium chloride
CAS Number: 138-24-9
MDL Number: MFCD00011790
Molecular Formula: C9H14ClN
Molecular Weight: 171.67
Purity/Analysis Method: 98.0% (HPLC,T)
Form: Crystal

Other Product Names / Synonyms: Phenyltrimethylammonium Chloride


Formula: C₉H₁₄ClN
MW: 171.67 g/mol
Melting Pt: 246…247 °C
MDL Number: MFCD00011790
CAS Number: 138-24-9
EINECS: 205-319-0
UN: 2811
ADR: 6.1,III
https://us-legacy.vwr.com/stibo/web/std.lang.all/09/75/28990975.jpg
Order Now
Specification Test Results
Purity/Analysis Method 98.0% (HPLC,T)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty