Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

o-Cresolphthalein complexone tetrasodium salt

Supplier: TCI America
Synonyms: o-Cresolphthalein Complexone sodium salt

P0344-1G
TCP0344-001GEA 142.1 USD InStock
TCP0344-001G
o-Cresolphthalein complexone tetrasodium salt
o-Cresolphthalein complexone tetrasodium salt
CAS Number: 62698-54-8
MDL Number: MFCD00066371
Form: Crystal
Color: Very Pale Yellow

Other Product Names / Synonyms: Cresolphthalein-bis(methyliminodiacetic Acid) Sodium Salt, o-Cresolphthalein Complexon Sodium Salt, Metal Phthalein Sodium Salt, Phthalein Purple Sodium Salt


Formula: C₃₂H₂₈N₂Na₄O₁₂
MDL Number: MFCD00066371
CAS Number: 62698-54-8
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty