Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Phthalocyanine ≥93.0% (by total nitrogen basis)

Supplier: TCI America
Synonyms: 29H,31H-Phthalocyanine Pigment Blue 16

P0355-25G
TCP0355-025GEA 118.76 USD InStock
TCP0355-025G
Phthalocyanine ≥93.0% (by total nitrogen basis)
Phthalocyanine
CAS Number: 574-93-6
MDL Number: MFCD00005085
Molecular Formula: C32H18N8
Molecular Weight: 514.55
Purity/Analysis Method: 93.0% (N)
Form: Crystal

Product Categories: Synthetic Organic Chemistry, Ligands, Materials Chemistry, Molecular Conductors, Porphyrin & Phthalocyanine Dyes (Functional Organic Dyes), Building Blocks for Phthalocyanines
Other Product Names / Synonyms: Pigment Blue 16


Formula: C₃₂H₁₈N₈
Melting Pt: 210 °C
Density: 1.447 g/cm³ (–113 °C)
MDL Number: MFCD00005085
CAS Number: 574-93-6
EINECS: 209-378-3
https://us-legacy.vwr.com/stibo/web/std.lang.all/10/82/28991082.jpg
Order Now
Specification Test Results
Purity/Analysis Method 93.0% (N)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty