Synonyms:
29H,31H-Phthalocyaninato-N29,N30,N31,N32 copper Pigment Blue 15
P0655-25G
TCP0655-025GEA
64.96
USD
InStock
TCP0655-025G
Copper(II) phthalocyanine
Copper (II) phthalocyanine
CAS Number: 147-14-8
MDL Number: MFCD00010719
Molecular Formula: C32H16CuN8
Molecular Weight: 576.08
Form: Crystal
Color: Deep Blue
Melting point (°C): 480
Lambda max.: 602 nm (CHCl3)
Other Product Names / Synonyms: Phthalocyanine Blue, Copper Phthalocyanine, CuPc