Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Pyrrole-2-carboxylic acid ≥98.0% (by titrimetric analysis)

Supplier: TCI America

P1270-1G P1270-5G
TCP1270-001GEA 39.08 USD InStock
TCP1270-001G TCP1270-005G
Pyrrole-2-carboxylic acid ≥98.0% (by titrimetric analysis)
Pyrrole-2-carboxylic acid
CAS Number: 634-97-9
MDL Number: MFCD00005219
Molecular Formula: C5H5NO2
Molecular Weight: 111.10
Purity/Analysis Method: 98.0% (T)
Form: Crystal
Color: Pale Gray
Melting point (°C): 208


Formula: C₅H₅NO₂
MW: 111.10 g/mol
Melting Pt: 208 °C
MDL Number: MFCD00005219
CAS Number: 634-97-9
EINECS: 211-221-9
https://us-legacy.vwr.com/stibo/web/std.lang.all/19/01/28991901.jpg
Order Now
Specification Test Results
Purity/Analysis Method 98.0% (T)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty