Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.

n-Heptane ≥99.0%

Supplier: TCI America
Danger

Q0037-100ML
TCQ0037-100MLEA 31.47 USD InStock
TCQ0037-100ML
n-Heptane ≥99.0%
n-Heptane
[Sequencing Solvent]
CAS Number: 142-82-5
MDL Number: MFCD00009544
Molecular Formula: C7H16
Molecular Weight: 100.21
Purity/Analysis Method: 99.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 98
Melting point (°C): -91
Flash Point (°C): -2
Specific Gravity (20/20): 0.68


Formula: H₃C(CH₂)₅CH₃
MW: 100.20 g/mol
Boiling Pt: 98 °C
Melting Pt: –91 °C
Density: 0.68 (20 °C)
Flash Pt: –2 °C
Storage Temperature: Ambient
MDL Number: MFCD00009544
CAS Number: 142-82-5
EINECS: 205-563-8
UN: 1206
ADR: 3,II
Merck Index: 13,04679
https://us-legacy.vwr.com/stibo/web/std.lang.all/36/95/28993695.jpg
Order Now
Specification Test Results
Purity/Analysis Method 99.0% (GC)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty