Synonyms:
EtOAc
, Acetidin
, Ethyl ethanoate
, Vinegar naphtha
, Ethyl acetic ester
, Acetic acid ethyl ester
, Acetic ether
, Acetoxyethane
Q0040-500ML
TCQ0040-500MLEA
39.08
USD
InStock
TCQ0040-500ML
Ethyl acetate ≥98.0%
Ethyl acetate
[Sequencing Solvent]
CAS Number: 141-78-6
MDL Number: MFCD00009171
Molecular Formula: C4H8O2
Molecular Weight: 88.11
Purity/Analysis Method: 98.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 77
Flash Point (°C): -3
Specific Gravity (20/20): 0.90
Other Product Names / Synonyms: Acetic Acid Ethyl Ester