Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Sesamol-(3,4-(methylenedioxy)phenol) ≥98.0%

Supplier: TCI America
Synonyms: Benzo[d][1,3]dioxol-5-olSesamol3,4-Methylenedioxyphenol

S0418-25G
TCS0418-025GEA 66.97 USD InStock
TCS0418-025G
Sesamol-(3,4-(methylenedioxy)phenol) ≥98.0%
Sesamol (3,4-(Methylenedioxy)phenol)
CAS Number: 533-31-3
MDL Number: MFCD00005827
Molecular Formula: C7H6O3
Molecular Weight: 138.12
Purity/Analysis Method: 98.0% (GC)
Form: Crystal
Color: White
Boiling point (°C): 113
Melting point (°C): 64
Flash Point (°C): 110

Product Categories: Bioscience, Antioxidants, Others (Phytochemicals)
Other Product Names / Synonyms: 5-Hydroxy-1,3-benzodioxole, 3,4-Methylenedioxyphenol


Formula: C₇H₆O₃
MW: 138.12 g/mol
Boiling Pt: 113 °C
Melting Pt: 64 °C
Flash Pt: 110 °C
MDL Number: MFCD00005827
CAS Number: 533-31-3
EINECS: 208-561-5
https://us-legacy.vwr.com/stibo/web/std.lang.all/41/08/28994108.jpg
Order Now
Specification Test Results
Purity/Analysis Method 98.0% (GC)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty