Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Tetrabutyltin ≥95.0%

Supplier: TCI America
Danger
Synonyms: Tetra-n-butyltin Tetrabutylstannane

T0058-500G T0058-25G
TCT0058-500GEA 436.45 USD InStock
TCT0058-500G TCT0058-025G
Tetrabutyltin ≥95.0%
Tetrabutyltin
CAS Number: 1461-25-2
MDL Number: MFCD00009422
Molecular Formula: C16H36Sn
Molecular Weight: 347.17
Purity/Analysis Method: 95.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 302
Melting point (°C): -97
Flash Point (°C): 107
Specific Gravity (20/20): 1.05


Formula: C₁₆H₃₆Sn
MW: 347.17 g/mol
Boiling Pt: 302 °C
Melting Pt: –97 °C
Density: 1.05 (20 °C)
Flash Pt: 107 °C
Storage Temperature: Ambient
MDL Number: MFCD00009422
CAS Number: 1461-25-2
EINECS: 215-960-8
UN: 2788
ADR: 6.1,III
https://us-legacy.vwr.com/stibo/web/std.lang.all/44/00/28994400.jpg
Order Now
Specification Test Results
Purity/Analysis Method 95.0% (GC)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty