Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

1,2,4,5-Tetrachlorobenzene ≥99.0%

Supplier: TCI America

T0060-500G T0060-25G
TCT0060-500GEA 159.28 USD InStock
TCT0060-500G TCT0060-025G
1,2,4,5-Tetrachlorobenzene ≥99.0%
1,2,4,5-Tetrachlorobenzene
CAS Number: 95-94-3
MDL Number: MFCD00000549
Molecular Formula: C6H2Cl4
Molecular Weight: 215.88
Purity/Analysis Method: 99.0% (GC)
Form: Crystal
Color: Very Pale Yellow
Boiling point (°C): 246
Melting point (°C): 140
Flash Point (°C): 155


Formula: C₆H₂Cl₄
MW: 215.89 g/mol
Boiling Pt: 246 °C
Melting Pt: 140 °C
Density: 1.732 g/cm³ (20 °C)
Flash Pt: 155 °C
Storage Temperature: Ambient
MDL Number: MFCD00000549
CAS Number: 95-94-3
EINECS: 202-466-2
UN: 3077
ADR: 9,III
https://us-legacy.vwr.com/stibo/web/std.lang.all/44/02/28994402.jpg
Order Now
Specification Test Results
Purity/Analysis Method 99.0% (GC)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty