Synonyms:
Ethanethioamide
T0187-100G
T0187-25G
T0187-500G
TCT0187-025GEA
28.54
USD
InStock
TCT0187-025G
TCT0187-100G
TCT0187-500G
Thioacetamide ≥98.0% (by GC, titration analysis)
Thioacetamide
CAS Number: 62-55-5
MDL Number: MFCD00008070
Molecular Formula: C2H5NS
Molecular Weight: 75.13
Purity/Analysis Method: 98.0% (GC,T)
Form: Crystal
Color: White
Melting point (°C): 112