Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Toluene ≥99.5%

Supplier: TCI America
Danger
Synonyms: MethacideBenzene, methylMethylbenzolToluolPhenylmethaneMethylbenzeneMethane, phenyl-

T0260-100ML
TCT0260-100MLEA 68.58 USD InStock
TCT0260-100ML
Toluene ≥99.5%
Toluene
[for Spectrophotometry]
CAS Number: 108-88-3
MDL Number: MFCD00008512
Molecular Formula: C7H8
Molecular Weight: 92.14
Purity/Analysis Method: 99.5% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 110
Melting point (°C): -95
Flash Point (°C): 5
Specific Gravity (20/20): 0.87


Formula: C₆H₅CH₃
MW: 92.14 g/mol
Boiling Pt: 110 °C
Melting Pt: –95 °C
Density: 0.87 (20 °C)
Flash Pt: 5 °C
Storage Temperature: Ambient
MDL Number: MFCD00008512
CAS Number: 108-88-3
EINECS: 203-625-9
UN: 1294
ADR: 3,II
REACH: 05-2114615130-69
Merck Index: 12,09667
https://us-legacy.vwr.com/stibo/web/std.lang.all/61/73/766173.jpg
Order Now
Specification Test Results
Purity/Analysis Method 99.5% (GC)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty